Product Details

SNP ID
rs1706810
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:45132088 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGAACCCAGCATCCTCTGGGCTCC[C/G]CCAAGGACAGGATCCATTTGATATT
Phenotype
MIM: 606758 MIM: 612771
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DUOX1 PubMed Links

Gene Details

Gene
DUOX1
Gene Name
dual oxidase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017434.4 Intron NP_059130.2
NM_175940.2 Intron NP_787954.1
XM_011521681.2 Intron XP_011519983.1
XM_011521682.1 Intron XP_011519984.1
Gene
DUOXA1
Gene Name
dual oxidase maturation factor 1
There are no transcripts associated with this gene.

View Full Product Details