Product Details

SNP ID
rs11178939
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:71766323 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGAAGTATCGTAGCATAGTAGTTA[C/T]AATTAATATCTGGAGCCAAACTGCC
Phenotype
MIM: 612398
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
RAB21 PubMed Links
Additional Information
For this assay, SNP(s) [rs74544711] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
RAB21
Gene Name
RAB21, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014999.3 Intron NP_055814.1

View Full Product Details