Product Details

SNP ID
rs919109
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:48598615 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACGCAGGATTGGAAAAAGTTAATT[C/G]TGTTTTCAGAAAAGTTAACCCCTGC
Phenotype
MIM: 142966 MIM: 142960 MIM: 142961 MIM: 142962
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HOXB-AS3 PubMed Links

Gene Details

Gene
HOXB-AS3
Gene Name
HOXB cluster antisense RNA 3
There are no transcripts associated with this gene.

Gene
HOXB3
Gene Name
homeobox B3
There are no transcripts associated with this gene.

Gene
HOXB5
Gene Name
homeobox B5
There are no transcripts associated with this gene.

Gene
HOXB6
Gene Name
homeobox B6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018952.4 Intron NP_061825.2
XM_005257283.3 Intron XP_005257340.1
XM_005257284.3 Intron XP_005257341.1
XM_011524727.2 Intron XP_011523029.1
Gene
HOXB7
Gene Name
homeobox B7
There are no transcripts associated with this gene.

View Full Product Details