Product Details

SNP ID
rs715239
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.4:99816878 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GCGAGAAGGTGTCAGGAGCAGCCCA[G/T]TTGTGTCTCTCTCTCTACCTCTGTG
Phenotype
MIM: 605768
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
DAPP1 PubMed Links
Additional Information
For this assay, SNP(s) [rs139586587] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
DAPP1
Gene Name
dual adaptor of phosphotyrosine and 3-phosphoinositides 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001306151.1 49 UTR 5 NP_001293080.1
NM_014395.2 49 UTR 5 NP_055210.2
XM_011531840.2 49 UTR 5 XP_011530142.1
XM_011531842.1 49 UTR 5 XP_011530144.1
XM_011531843.2 49 Intron XP_011530145.1
XM_017008023.1 49 UTR 5 XP_016863512.1
XM_017008024.1 49 UTR 5 XP_016863513.1

View Full Product Details