Product Details
- SNP ID
-
rs11941041
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:20704215 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGAGTTTCTAACAGTTTATTTTGAC[A/G]ATTTGAATGGAAAAAAAGTTGACTT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PACRGL
PubMed Links
Gene Details
- Gene
- PACRGL
- Gene Name
- PARK2 coregulated like
View Full Product Details