Product Details

SNP ID
rs2077543
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:41840002 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGGCAGGCGTCATGATGGAGATAT[C/G]ATGTCTCCTCTTGTTGATTCAGGAT
Phenotype
MIM: 606088 MIM: 605916
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
JMJD7 PubMed Links

Gene Details

Gene
JMJD7
Gene Name
jumonji domain containing 7
There are no transcripts associated with this gene.

Gene
JMJD7-PLA2G4B
Gene Name
JMJD7-PLA2G4B readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198588.1 Intron NP_001185517.1
NM_005090.3 Intron NP_005081.1
Gene
PLA2G4B
Gene Name
phospholipase A2 group IVB
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114633.1 Intron NP_001108105.1
Gene
SPTBN5
Gene Name
spectrin beta, non-erythrocytic 5
There are no transcripts associated with this gene.

View Full Product Details