Product Details

SNP ID
rs4737229
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.8:66436672 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GGACTTCGGGAGAGATGCCCAAGCC[A/G]GAAAGATAAATGTGGAAGTCATCAG
Phenotype
MIM: 611083
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADHFE1 PubMed Links

Gene Details

Gene
ADHFE1
Gene Name
alcohol dehydrogenase, iron containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144650.2 Intron NP_653251.2
Gene
RRS1
Gene Name
ribosome biogenesis regulator homolog
There are no transcripts associated with this gene.

Gene
RRS1-AS1
Gene Name
RRS1 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details