Product Details

SNP ID
rs11878667
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54537218 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGCCCACTGGATCACAGAGCCACA[C/T]GTGAGGGGCGTCCCGCTGGGCACAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIR3DX1 PubMed Links

Gene Details

Gene
KIR3DX1
Gene Name
killer cell immunoglobulin like receptor, three Ig domains X1
There are no transcripts associated with this gene.

View Full Product Details