Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_000141.4 | 3659 | UTR 3 | NP_000132.3 | ||
NM_001144913.1 | 3659 | Intron | NP_001138385.1 | ||
NM_001144914.1 | 3659 | UTR 3 | NP_001138386.1 | ||
NM_001144915.1 | 3659 | Silent Mutation | CTG,TTG | L703L | NP_001138387.1 |
NM_001144916.1 | 3659 | UTR 3 | NP_001138388.1 | ||
NM_001144917.1 | 3659 | UTR 3 | NP_001138389.1 | ||
NM_001144918.1 | 3659 | UTR 3 | NP_001138390.1 | ||
NM_001144919.1 | 3659 | Intron | NP_001138391.1 | ||
NM_001320654.1 | 3659 | UTR 3 | NP_001307583.1 | ||
NM_001320658.1 | 3659 | UTR 3 | NP_001307587.1 | ||
NM_022970.3 | 3659 | UTR 3 | NP_075259.4 | ||
NM_023029.2 | 3659 | UTR 3 | NP_075418.1 | ||
XM_006717708.2 | 3659 | UTR 3 | XP_006717771.1 | ||
XM_006717710.3 | 3659 | UTR 3 | XP_006717773.1 | ||
XM_006717711.2 | 3659 | UTR 3 | XP_006717774.1 | ||
XM_006717712.2 | 3659 | UTR 3 | XP_006717775.1 | ||
XM_017015920.1 | 3659 | UTR 3 | XP_016871409.1 | ||
XM_017015921.1 | 3659 | UTR 3 | XP_016871410.1 | ||
XM_017015922.1 | 3659 | UTR 3 | XP_016871411.1 | ||
XM_017015923.1 | 3659 | UTR 3 | XP_016871412.1 | ||
XM_017015924.1 | 3659 | UTR 3 | XP_016871413.1 | ||
XM_017015925.1 | 3659 | UTR 3 | XP_016871414.1 |