Product Details

SNP ID
rs3731993
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:170993819 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCTTGAAGAAGGGGGTGTAGGGGA[T/G]GCTGTCAGCCCAGCCATGTGTAGGT
Phenotype
MIM: 608438
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
TLK1 PubMed Links

Gene Details

Gene
TLK1
Gene Name
tousled like kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136554.1 2843 Silent Mutation GCA,GCC A706A NP_001130026.1
NM_001136555.1 2843 Silent Mutation GCA,GCC A658A NP_001130027.1
NM_012290.4 2843 Silent Mutation GCA,GCC A754A NP_036422.3
XM_011512238.2 2843 Silent Mutation GCA,GCC A691A XP_011510540.1
XM_017005421.1 2843 Silent Mutation GCA,GCC A537A XP_016860910.1
XM_017005422.1 2843 Silent Mutation GCA,GCC A537A XP_016860911.1

View Full Product Details