Product Details
- SNP ID
-
rs12824458
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:49234036 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAAGGCTCTCCCACTGGATGACGGT[C/G]ACGCGCAATAAGCAGATGATTGCAG
- Phenotype
-
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
TUBA1C
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114472800] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TUBA1C
- Gene Name
- tubulin alpha 1c
View Full Product Details