Product Details

SNP ID
rs4479364
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:40318381 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCCCCTCTCTGTGCCTCTGTAAAG[C/T]GCAACAATAAGAGAATCTCCCTGGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C19orf47 PubMed Links

Gene Details

Gene
C19orf47
Gene Name
chromosome 19 open reading frame 47
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256440.1 4106 Intron NP_001243369.1
NM_001256441.1 4106 Intron NP_001243370.1
XM_005258520.2 4106 Intron XP_005258577.1
XM_011526460.2 4106 Intron XP_011524762.1
XM_017026291.1 4106 Intron XP_016881780.1
XM_017026292.1 4106 UTR 3 XP_016881781.1
XM_017026293.1 4106 UTR 3 XP_016881782.1
XM_017026294.1 4106 Intron XP_016881783.1
XM_017026295.1 4106 Intron XP_016881784.1

View Full Product Details