Product Details

SNP ID
rs17831104
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:24619617 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TACAGACAAGGTAAAAATCCTGGGA[C/T]GTAGGTGGAAGGAAAAACTTCACAG
Phenotype
MIM: 610739
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TNRC6A PubMed Links

Gene Details

Gene
TNRC6A
Gene Name
trinucleotide repeat containing 6A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014494.2 Intron NP_055309.2
XM_005255254.3 Intron XP_005255311.1
XM_005255257.4 Intron XP_005255314.1
XM_017023143.1 Intron XP_016878632.1
XM_017023144.1 Intron XP_016878633.1
XM_017023145.1 Intron XP_016878634.1
XM_017023146.1 Intron XP_016878635.1
XM_017023147.1 Intron XP_016878636.1
XM_017023148.1 Intron XP_016878637.1
XM_017023149.1 Intron XP_016878638.1
XM_017023150.1 Intron XP_016878639.1
XM_017023151.1 Intron XP_016878640.1
XM_017023152.1 Intron XP_016878641.1
XM_017023153.1 Intron XP_016878642.1
XM_017023154.1 Intron XP_016878643.1

View Full Product Details