Product Details

SNP ID
rs1045764
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:53108220 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTTGTGATGAAGGGTTTGCTGTACT[C/T]GTAAGGATTCTCTCAAGAATGAAAT
Phenotype
MIM: 600398
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF160 PubMed Links

Gene Details

Gene
ZNF160
Gene Name
zinc finger protein 160
There are no transcripts associated with this gene.

Gene
ZNF415
Gene Name
zinc finger protein 415
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136038.2 2511 UTR 3 NP_001129510.2
NM_001164309.1 2511 UTR 3 NP_001157781.1
NM_018355.3 2511 UTR 3 NP_060825.2
XM_006723267.1 2511 UTR 3 XP_006723330.1
XM_006723268.1 2511 UTR 3 XP_006723331.1
XM_006723272.2 2511 UTR 3 XP_006723335.1
XM_006723273.2 2511 UTR 3 XP_006723336.1
XM_006723274.2 2511 UTR 3 XP_006723337.1
XM_006723277.2 2511 UTR 3 XP_006723340.1
XM_006723279.1 2511 UTR 3 XP_006723342.1
XM_006723280.2 2511 UTR 3 XP_006723343.1
XM_006723281.2 2511 UTR 3 XP_006723344.1
XM_006723282.1 2511 UTR 3 XP_006723345.1
XM_011527100.1 2511 UTR 3 XP_011525402.1
XM_011527101.2 2511 UTR 3 XP_011525403.1
XM_011527103.1 2511 UTR 3 XP_011525405.1
XM_011527104.1 2511 UTR 3 XP_011525406.1
XM_017026952.1 2511 UTR 3 XP_016882441.1
XM_017026953.1 2511 UTR 3 XP_016882442.1
XM_017026954.1 2511 UTR 3 XP_016882443.1
XM_017026955.1 2511 UTR 3 XP_016882444.1
XM_017026956.1 2511 UTR 3 XP_016882445.1
XM_017026957.1 2511 UTR 3 XP_016882446.1
XM_017026958.1 2511 UTR 3 XP_016882447.1
XM_017026959.1 2511 UTR 3 XP_016882448.1
XM_017026960.1 2511 UTR 3 XP_016882449.1
XM_017026961.1 2511 UTR 3 XP_016882450.1
XM_017026962.1 2511 UTR 3 XP_016882451.1
XM_017026963.1 2511 UTR 3 XP_016882452.1
XM_017026964.1 2511 UTR 3 XP_016882453.1
XM_017026965.1 2511 UTR 3 XP_016882454.1
XM_017026966.1 2511 UTR 3 XP_016882455.1
XM_017026967.1 2511 UTR 3 XP_016882456.1
XM_017026968.1 2511 UTR 3 XP_016882457.1
XM_017026969.1 2511 UTR 3 XP_016882458.1
XM_017026970.1 2511 UTR 3 XP_016882459.1
XM_017026971.1 2511 UTR 3 XP_016882460.1
XM_017026972.1 2511 UTR 3 XP_016882461.1
XM_017026973.1 2511 UTR 3 XP_016882462.1
XM_017026974.1 2511 UTR 3 XP_016882463.1

View Full Product Details