Product Details

SNP ID
rs12609357
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:47996052 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GATATTGGTGCCAATGTCTTATCTA[C/T]AGCTGTTTGAAGACAGCTATCATAT
Phenotype
MIM: 612213 MIM: 607443
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BSPH1 PubMed Links

Gene Details

Gene
BSPH1
Gene Name
binder of sperm protein homolog 1
There are no transcripts associated with this gene.

Gene
ELSPBP1
Gene Name
epididymal sperm binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022142.4 Intron NP_071425.3
XM_017027130.1 Intron XP_016882619.1

View Full Product Details