Product Details

SNP ID
rs475002
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:7921753 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GATAACAGGGCCACTGGAAGAAGCC[C/G]TGGCAGTGGCTTTCTCGCAGGTACC
Phenotype
MIM: 600135 MIM: 603014 MIM: 605076 MIM: 605058
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CTXN1 PubMed Links

Gene Details

Gene
CTXN1
Gene Name
cortexin 1
There are no transcripts associated with this gene.

Gene
MAP2K7
Gene Name
mitogen-activated protein kinase kinase 7
There are no transcripts associated with this gene.

Gene
SNAPC2
Gene Name
small nuclear RNA activating complex polypeptide 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003083.3 410 Missense Mutation CTG,GTG L118V NP_003074.1
Gene
TGFBR3L
Gene Name
transforming growth factor beta receptor 3 like
There are no transcripts associated with this gene.

Gene
TIMM44
Gene Name
translocase of inner mitochondrial membrane 44
There are no transcripts associated with this gene.

View Full Product Details