Product Details

SNP ID
rs1864074
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:40598954 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATAACGTATTTTTTCAATAAAATC[G/T]TACGCCAAAATCTCTTCATAAGACA
Phenotype
MIM: 604710
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LTBP4 PubMed Links

Gene Details

Gene
LTBP4
Gene Name
latent transforming growth factor beta binding protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042544.1 Intron NP_001036009.1
NM_001042545.1 Intron NP_001036010.1
NM_003573.2 Intron NP_003564.2
XM_011527376.2 Intron XP_011525678.1
XM_011527377.2 Intron XP_011525679.1
XM_011527378.2 Intron XP_011525680.1
XM_011527379.1 Intron XP_011525681.1
XM_011527380.2 Intron XP_011525682.1
XM_011527381.2 Intron XP_011525683.1
XM_011527382.2 Intron XP_011525684.1
XM_011527383.2 Intron XP_011525685.1
XM_011527384.2 Intron XP_011525686.1
XM_011527385.2 Intron XP_011525687.1
XM_011527386.2 Intron XP_011525688.1
XM_011527387.1 Intron XP_011525689.1
XM_017027352.1 Intron XP_016882841.1
XM_017027353.1 Intron XP_016882842.1
XM_017027354.1 Intron XP_016882843.1
Gene
SHKBP1
Gene Name
SH3KBP1 binding protein 1
There are no transcripts associated with this gene.

View Full Product Details