Product Details

SNP ID
rs7257939
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12618282 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTGCCTCCTTTGAACATTTAATGA[G/T]TATTAATGGAATTACACTTAAAGGG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF490 PubMed Links

Gene Details

Gene
ZNF490
Gene Name
zinc finger protein 490
There are no transcripts associated with this gene.

Gene
ZNF791
Gene Name
zinc finger protein 791
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153358.2 Intron NP_699189.2
XM_017026412.1 Intron XP_016881901.1

View Full Product Details