Product Details

SNP ID
rs11949556
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:150850239 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTCACATACGTAATTTAAAAAAAT[A/C]TGTATGGCAGTAAAAGATGTGTGCT
Phenotype
MIM: 608212
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
IRGM PubMed Links

Gene Details

Gene
IRGM
Gene Name
immunity related GTPase M
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145805.1 Intron NP_001139277.1
XM_011537641.2 Intron XP_011535943.1

View Full Product Details