Product Details

SNP ID
rs1046224
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.5:38475405 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CCTTATACGATTAAACATGATTTTA[A/G]GTACAGCACATCACAACTGTTTATT
Phenotype
MIM: 151443
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EGFLAM PubMed Links

Gene Details

Gene
EGFLAM
Gene Name
EGF like, fibronectin type III and laminin G domains
There are no transcripts associated with this gene.

Gene
LIFR
Gene Name
leukemia inhibitory factor receptor alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127671.1 9661 UTR 3 NP_001121143.1
NM_002310.5 9661 UTR 3 NP_002301.1
XM_011514040.2 9661 Intron XP_011512342.1
XM_011514042.2 9661 Intron XP_011512344.1
XM_017009462.1 9661 UTR 3 XP_016864951.1
XM_017009463.1 9661 Intron XP_016864952.1

View Full Product Details