Product Details

SNP ID
rs11880385
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:17298480 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTACAGGATTCCGATGAGCGCTTC[C/G]TGAATGCACAGTGTTCTCCAGGGCA
Phenotype
MIM: 611840
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ABHD8 PubMed Links

Gene Details

Gene
ABHD8
Gene Name
abhydrolase domain containing 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024527.4 Intron NP_078803.4
Gene
MRPL34
Gene Name
mitochondrial ribosomal protein L34
There are no transcripts associated with this gene.

View Full Product Details