Product Details

SNP ID
rs7250522
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:49363128 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
ACTGGAGACGGCTGGCGGCTGTCCC[A/G]GGACGGGGAAGAGAGCTTTGGTGCC
Phenotype
MIM: 605418 MIM: 601729
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DKKL1 PubMed Links

Gene Details

Gene
DKKL1
Gene Name
dickkopf like acrosomal protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001197301.1 Intron NP_001184230.1
NM_001197302.1 Intron NP_001184231.1
NM_014419.3 Intron NP_055234.1
XM_006723142.2 Intron XP_006723205.1
XM_011526724.1 Intron XP_011525026.1
XM_011526725.1 Intron XP_011525027.1
XM_011526726.2 Intron XP_011525028.1
XM_011526727.2 Intron XP_011525029.1
XM_011526728.2 Intron XP_011525030.1
XM_011526729.1 Intron XP_011525031.1
XM_017026593.1 Intron XP_016882082.1
Gene
LOC101928295
Gene Name
uncharacterized LOC101928295
There are no transcripts associated with this gene.

Gene
TEAD2
Gene Name
TEA domain transcription factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256658.1 Intron NP_001243587.1
NM_001256659.1 Intron NP_001243588.1
NM_001256660.1 Intron NP_001243589.1
NM_001256661.1 Intron NP_001243590.1
NM_001256662.1 Intron NP_001243591.1
NM_003598.1 Intron NP_003589.1
XM_005259334.3 Intron XP_005259391.1
XM_006723424.1 Intron XP_006723487.1
XM_006723428.2 Intron XP_006723491.1
XM_006723429.2 Intron XP_006723492.1
XM_011527398.1 Intron XP_011525700.1
XM_011527399.2 Intron XP_011525701.1
XM_011527400.2 Intron XP_011525702.1
XM_011527401.1 Intron XP_011525703.1
XM_011527402.2 Intron XP_011525704.1
XM_011527403.1 Intron XP_011525705.1
XM_011527404.1 Intron XP_011525706.1
XM_011527405.2 Intron XP_011525707.1
XM_011527406.2 Intron XP_011525708.1

View Full Product Details