Product Details

SNP ID
rs3823036
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:98836656 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACATGCTGCTGTGTTTATTTATTG[C/T]GGATTCCCATCAGGAAAGAGGAAAA
Phenotype
MIM: 600494
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
POU3F2 PubMed Links

Gene Details

Gene
POU3F2
Gene Name
POU class 3 homeobox 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005604.3 1953 UTR 3 NP_005595.2

View Full Product Details