Product Details
- SNP ID
-
rs2422764
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:2387422 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- GTCACTGTTGAACACTGGTCCTGCA[C/G]AGTGCTCACTCACCACAGCAGGGGC
- Phenotype
-
MIM: 613900
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TGM6
PubMed Links
Gene Details
- Gene
- TGM6
- Gene Name
- transglutaminase 6
View Full Product Details