Product Details

SNP ID
rs4732847
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:26741644 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACCTGTGCCATGGCAGGGTGTGT[A/C]CTAAAGCATTCTATAGTCCTGGGTC
Phenotype
MIM: 104221
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ADRA1A PubMed Links

Gene Details

Gene
ADRA1A
Gene Name
adrenoceptor alpha 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000680.3 12978 Intron NP_000671.2
NM_001322502.1 12978 Intron NP_001309431.1
NM_001322503.1 12978 Intron NP_001309432.1
NM_001322504.1 12978 Intron NP_001309433.1
NM_033302.3 12978 Intron NP_150645.2
NM_033303.4 12978 Intron NP_150646.3
NM_033304.3 12978 Intron NP_150647.2
XM_006716292.3 12978 UTR 3 XP_006716355.1
XM_006716293.3 12978 Intron XP_006716356.1
XM_011544411.2 12978 Intron XP_011542713.1
XM_011544412.2 12978 Intron XP_011542714.1
XM_017013094.1 12978 UTR 3 XP_016868583.1
XM_017013095.1 12978 UTR 3 XP_016868584.1
XM_017013096.1 12978 Intron XP_016868585.1

View Full Product Details