Product Details

SNP ID
rs937352
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:26741198 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGATCCTATTCATGCCTCTCTAAC[C/G]TTTTACCCCACTCATTCTCTCTCAC
Phenotype
MIM: 104221
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ADRA1A PubMed Links

Gene Details

Gene
ADRA1A
Gene Name
adrenoceptor alpha 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000680.3 13424 Intron NP_000671.2
NM_001322502.1 13424 Intron NP_001309431.1
NM_001322503.1 13424 Intron NP_001309432.1
NM_001322504.1 13424 Intron NP_001309433.1
NM_033302.3 13424 Intron NP_150645.2
NM_033303.4 13424 Intron NP_150646.3
NM_033304.3 13424 Intron NP_150647.2
XM_006716292.3 13424 UTR 3 XP_006716355.1
XM_006716293.3 13424 Intron XP_006716356.1
XM_011544411.2 13424 Intron XP_011542713.1
XM_011544412.2 13424 Intron XP_011542714.1
XM_017013094.1 13424 UTR 3 XP_016868583.1
XM_017013095.1 13424 UTR 3 XP_016868584.1
XM_017013096.1 13424 Intron XP_016868585.1

View Full Product Details