Product Details

SNP ID
rs35177469
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:48145922 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCACAGATTCTTAAACCAGAGTC[C/T]GTGAACCCCTGGCCTCTCTGTGGAT
Phenotype
MIM: 604969
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SKAP1 PubMed Links

Gene Details

Gene
SKAP1
Gene Name
src kinase associated phosphoprotein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001075099.1 Intron NP_001068567.1
NM_003726.3 Intron NP_003717.3
XM_005257755.3 Intron XP_005257812.1
XM_017025257.1 Intron XP_016880746.1
XM_017025258.1 Intron XP_016880747.1
XM_017025259.1 Intron XP_016880748.1
XM_017025260.1 Intron XP_016880749.1

View Full Product Details