Product Details

SNP ID
rs2240921
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.3:52796748 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
CTCTCTTTCCCTGCAGGACCATCGA[C/T]GGTGTTACCTACCCTGGGAATGTCA
Phenotype
MIM: 147270 MIM: 146650
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ITIH1 PubMed Links

Gene Details

Gene
ITIH1
Gene Name
inter-alpha-trypsin inhibitor heavy chain 1
There are no transcripts associated with this gene.

Gene
ITIH3
Gene Name
inter-alpha-trypsin inhibitor heavy chain 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002217.3 327 Silent Mutation GAC,GAT D97D NP_002208.3
XM_005265105.4 327 Silent Mutation GAC,GAT D97D XP_005265162.1
XM_017006356.1 327 UTR 5 XP_016861845.1

View Full Product Details