Product Details

SNP ID
rs34358803
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:47973488 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGGAATTTTGGTGATGTGAATGG[G/T]AATGCTCTAATTTGGGTGCTTCTCA
Phenotype
MIM: 608202
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CDK5RAP3 PubMed Links

Gene Details

Gene
CDK5RAP3
Gene Name
CDK5 regulatory subunit associated protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278197.1 Intron NP_001265126.1
NM_001278198.1 Intron NP_001265127.1
NM_001278216.1 Intron NP_001265145.1
NM_001278217.1 Intron NP_001265146.1
NM_176096.2 Intron NP_788276.1
XM_011525297.1 Intron XP_011523599.1
XM_017025164.1 Intron XP_016880653.1
XM_017025165.1 Intron XP_016880654.1
XM_017025166.1 Intron XP_016880655.1

View Full Product Details