Product Details

SNP ID
rs1150705
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:28226008 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGGAACACCCCCATGGGTTTCAGG[G/T]GCTTCCTGCGTCTGTGGAGAAAACT
Phenotype
MIM: 602246
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TOB2P1 PubMed Links

Gene Details

Gene
TOB2P1
Gene Name
transducer of ERBB2, 2 pseudogene 1
There are no transcripts associated with this gene.

Gene
ZSCAN9
Gene Name
zinc finger and SCAN domain containing 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199479.1 Intron NP_001186408.1
NM_001199480.1 Intron NP_001186409.1
NM_006299.4 Intron NP_006290.1
XM_011514875.2 Intron XP_011513177.1
XM_011514876.2 Intron XP_011513178.1
XM_011514878.2 Intron XP_011513180.1
XM_017011267.1 Intron XP_016866756.1

View Full Product Details