Product Details

SNP ID
rs8050532
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:5072767 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTAAGATTCAGGCCCTATCTCTGTG[G/T]GCCTCCTAGAGCAATCTGAACAAGA
Phenotype
MIM: 605907
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ALG1 PubMed Links

Gene Details

Gene
ALG1
Gene Name
ALG1, chitobiosyldiphosphodolichol beta-mannosyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019109.4 Intron NP_061982.3
XM_011522565.1 Intron XP_011520867.1
XM_017023457.1 Intron XP_016878946.1
XM_017023458.1 Intron XP_016878947.1
Gene
C16orf89
Gene Name
chromosome 16 open reading frame 89
There are no transcripts associated with this gene.

View Full Product Details