Product Details

SNP ID
rs932019
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:132263677 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGCCGTGTTCACTGCTGAGGGGA[C/T]TTCCATGACCCCCTGTGGAAGTACG
Phenotype
MIM: 603422
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PDLIM4 PubMed Links

Gene Details

Gene
PDLIM4
Gene Name
PDZ and LIM domain 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001131027.1 Intron NP_001124499.1
NM_003687.3 Intron NP_003678.2
XM_017010002.1 Intron XP_016865491.1

View Full Product Details