Product Details

SNP ID
rs1064826
Assay Type
Functionally tested
NCBI dbSNP Submissions
62
Location
Chr.1:8352929 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTTCTTTGGAAAAATGGATGCCA[A/G]AGGAGGAGATAAAGAAGGTTTAACT
Phenotype
MIM: 605226 MIM: 605763
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RERE PubMed Links

Gene Details

Gene
RERE
Gene Name
arginine-glutamic acid dipeptide repeats
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042681.1 7409 UTR 3 NP_001036146.1
NM_001042682.1 7409 UTR 3 NP_001036147.1
NM_012102.3 7409 UTR 3 NP_036234.3
XM_005263464.2 7409 UTR 3 XP_005263521.1
XM_005263466.1 7409 UTR 3 XP_005263523.1
XM_011541510.1 7409 UTR 3 XP_011539812.1
XM_011541511.1 7409 Intron XP_011539813.1
XM_017001358.1 7409 UTR 3 XP_016856847.1
XM_017001359.1 7409 UTR 3 XP_016856848.1
Gene
SLC45A1
Gene Name
solute carrier family 45 member 1
There are no transcripts associated with this gene.

View Full Product Details