Product Details

SNP ID
rs14089
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:2997792 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGGCGGCTGCTAGGATGTCTGTCC[C/T]GGCTGCTGATTCCCCTGGGAGTCTG
Phenotype
MIM: 601041 MIM: 612399
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TLE2 PubMed Links

Gene Details

Gene
TLE2
Gene Name
transducin like enhancer of split 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144761.1 2588 UTR 3 NP_001138233.1
NM_001144762.1 2588 UTR 3 NP_001138234.1
NM_001300846.1 2588 UTR 3 NP_001287775.1
NM_003260.4 2588 UTR 3 NP_003251.2
XM_006722864.2 2588 UTR 3 XP_006722927.1
XM_011528230.1 2588 UTR 3 XP_011526532.1
Gene
TLE6
Gene Name
transducin like enhancer of split 6
There are no transcripts associated with this gene.

View Full Product Details