Product Details

SNP ID
rs308038
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:3098145 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTCACTACTCAGTGTATGCATGA[A/G]TGTGAATGCATGTATTGTTTTATAC
Phenotype
MIM: 139313
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GNA11 PubMed Links

Gene Details

Gene
GNA11
Gene Name
G protein subunit alpha 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002067.4 Intron NP_002058.2

View Full Product Details