Product Details

SNP ID
rs735119
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:50190317 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCGGCCCAGGAAAGCTTCTCTGCCC[G/T]CCCTGACAGGAGACCTGTGCACGCC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TRABD PubMed Links

Gene Details

Gene
TRABD
Gene Name
TraB domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320484.1 Intron NP_001307413.1
NM_001320485.1 Intron NP_001307414.1
NM_001320487.1 Intron NP_001307416.1
NM_001320488.1 Intron NP_001307417.1
NM_025204.3 Intron NP_079480.2
XM_006724422.3 Intron XP_006724485.1
XM_006724423.3 Intron XP_006724486.1
XM_011530712.2 Intron XP_011529014.2
XM_011530713.2 Intron XP_011529015.2
XM_011530715.2 Intron XP_011529017.2
XM_011530716.2 Intron XP_011529018.1
XM_011530717.2 Intron XP_011529019.1
XM_011530718.2 Intron XP_011529020.1
XM_017028943.1 Intron XP_016884432.1
XM_017028944.1 Intron XP_016884433.1

View Full Product Details