Product Details
- SNP ID
-
rs4838859
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:50174773 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CAGTCGCTGCGTGTGGGATGGGCCC[A/G]TGGATTTGGGCTCAGCAGCCCCTCT
- Phenotype
-
MIM: 608421
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PANX2
PubMed Links
Gene Details
- Gene
- PANX2
- Gene Name
- pannexin 2
View Full Product Details