Product Details
- SNP ID
-
rs35236393
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:50174159 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGGCCAGCAGGCAGGAGGAGGCTG[C/T]GCAGAGCCAGGGGTGTCCTGGAGAG
- Phenotype
-
MIM: 608421
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PANX2
PubMed Links
Gene Details
- Gene
- PANX2
- Gene Name
- pannexin 2
View Full Product Details