Product Details
- SNP ID
-
rs4270426
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:31307826 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- ATGACTGAATAAGATACCCACTTTA[A/G]GCAAACACTAACTACAACTCAAGCC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DEFB116
PubMed Links
Gene Details
- Gene
- DEFB116
- Gene Name
- defensin beta 116
View Full Product Details