Product Details

SNP ID
rs627222
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:6088824 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGAAGCCTGAAGTGTTCCAAGCAT[G/A]GAGATGATGGACGTGGTATGGGACA
Phenotype
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
USP42 PubMed Links

Gene Details

Gene
USP42
Gene Name
ubiquitin specific peptidase 42
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032172.2 466 Intron NP_115548.1
XM_005249883.4 466 Intron XP_005249940.1
XM_006715790.1 466 Intron XP_006715853.1
XM_006715791.2 466 UTR 5 XP_006715854.1
XM_011515573.1 466 UTR 5 XP_011513875.1
XM_011515574.1 466 Intron XP_011513876.1
XM_011515577.1 466 Intron XP_011513879.1
XM_011515578.1 466 Intron XP_011513880.1

View Full Product Details