Product Details

SNP ID
rs6834890
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:13338847 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TACTCTGATAGCAAGATTGGATGTT[A/G]TATATTATGAGGCTTTTTGTTTATT
Phenotype
MIM: 612994
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HSP90AB2P PubMed Links

Gene Details

Gene
HSP90AB2P
Gene Name
heat shock protein 90 alpha family class B member 2, pseudogene
There are no transcripts associated with this gene.

Gene
RAB28
Gene Name
RAB28, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017979.2 Intron NP_001017979.1
NM_001159601.1 Intron NP_001153073.1
NM_004249.3 Intron NP_004240.2
XM_005248218.4 Intron XP_005248275.1
XM_011513911.2 Intron XP_011512213.1
XM_011513912.2 Intron XP_011512214.1
XM_017008850.1 Intron XP_016864339.1

View Full Product Details