Product Details

SNP ID
rs12581653
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:45064723 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGTTTTTGCAACTATGGATTCATT[A/C]CCCCTGGTCTACTGTGGAGCCAATG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RACGAP1P PubMed Links

Gene Details

Gene
RACGAP1P
Gene Name
Rac GTPase activating protein 1 pseudogene
There are no transcripts associated with this gene.

View Full Product Details