Product Details

SNP ID
rs2844676
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30988438 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACACACGACAAAGAGAAGCTGTGC[G/T]TGCCCCGGGGTGGGTATCTAGCTCT
Phenotype
MIM: 616991
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MUC21 PubMed Links

Gene Details

Gene
MUC21
Gene Name
mucin 21, cell surface associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010909.3 2196 UTR 3 NP_001010909.2
NM_001322370.1 2196 Intron NP_001309299.1
NM_001322371.1 2196 Intron NP_001309300.1

View Full Product Details