Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001007024.1 | 33 | UTR 5 | NP_001007025.1 | ||
NM_001007025.1 | 33 | Missense Mutation | ACA,GCA | T3A | NP_001007026.1 |
NM_004871.2 | 33 | Missense Mutation | ACA,GCA | T3A | NP_004862.1 |
XM_005258070.4 | 33 | Intron | XP_005258127.1 | ||
XM_005258071.4 | 33 | Intron | XP_005258128.1 | ||
XM_005258072.2 | 33 | UTR 5 | XP_005258129.1 | ||
XM_017025374.1 | 33 | Intron | XP_016880863.1 | ||
XM_017025375.1 | 33 | Missense Mutation | ACA,GCA | T3A | XP_016880864.1 |
XM_017025376.1 | 33 | Missense Mutation | ACA,GCA | T3A | XP_016880865.1 |
XM_017025377.1 | 33 | Intron | XP_016880866.1 |