Product Details

SNP ID
rs2040533
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:31283124 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCGGCCTGGCCCATAAGGGGGGTG[C/G]TGCCCTCCTGAGGGTCAACTGGAGT
Phenotype
MIM: 601988
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LIMK2 PubMed Links

Gene Details

Gene
LIMK2
Gene Name
LIM domain kinase 2
There are no transcripts associated with this gene.

Gene
PIK3IP1
Gene Name
phosphoinositide-3-kinase interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135911.1 942 UTR 3 NP_001129383.1
NM_052880.4 942 Missense Mutation ACC,AGC T251S NP_443112.2

View Full Product Details