Product Details

SNP ID
rs11605738
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:64245934 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTGAGTCACCGTCTGAACAACCAG[A/G]GAGTCATGGGGGTGGAGAAGCAGAG
Phenotype
MIM: 186946 MIM: 600230 MIM: 601140 MIM: 601398
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FKBP2 PubMed Links

Gene Details

Gene
FKBP2
Gene Name
FK506 binding protein 2
There are no transcripts associated with this gene.

Gene
LOC105369340
Gene Name
uncharacterized LOC105369340
There are no transcripts associated with this gene.

Gene
PLCB3
Gene Name
phospholipase C beta 3
There are no transcripts associated with this gene.

Gene
PPP1R14B
Gene Name
protein phosphatase 1 regulatory inhibitor subunit 14B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138689.2 Intron NP_619634.1
Gene
VEGFB
Gene Name
vascular endothelial growth factor B
There are no transcripts associated with this gene.

View Full Product Details