Product Details

SNP ID
rs580192
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:23705437 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTCCCACACACACCCAGACTGGTC[C/T]GAAATGTTTCATTTCAGTGGGGTGA
Phenotype
MIM: 300862
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACOT9 PubMed Links

Gene Details

Gene
ACOT9
Gene Name
acyl-CoA thioesterase 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033583.2 Intron NP_001028755.2
NM_001037171.1 Intron NP_001032248.1
XM_005274472.2 Intron XP_005274529.1

View Full Product Details