Product Details

SNP ID
rs9607973
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.22:42872496 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ATTTTTTGGTCCATTTTTCAGAATT[A/G]GATGCAGTTTTCCCCCTGGTTTAGC
Phenotype
MIM: 604960
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PACSIN2 PubMed Links

Gene Details

Gene
PACSIN2
Gene Name
protein kinase C and casein kinase substrate in neurons 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184970.1 Intron NP_001171899.1
NM_001184971.1 Intron NP_001171900.1
NM_007229.3 Intron NP_009160.2
XM_005261319.3 Intron XP_005261376.1
XM_011529846.1 Intron XP_011528148.1
XM_011529847.2 Intron XP_011528149.1
XM_011529848.2 Intron XP_011528150.1
XM_011529849.2 Intron XP_011528151.1
XM_011529850.2 Intron XP_011528152.1
XM_011529851.2 Intron XP_011528153.1
XM_011529852.1 Intron XP_011528154.1
XM_011529853.1 Intron XP_011528155.1
XM_017028562.1 Intron XP_016884051.1
XM_017028563.1 Intron XP_016884052.1
XM_017028564.1 Intron XP_016884053.1
XM_017028565.1 Intron XP_016884054.1
XM_017028566.1 Intron XP_016884055.1
XM_017028567.1 Intron XP_016884056.1
XM_017028568.1 Intron XP_016884057.1

View Full Product Details