Product Details

SNP ID
rs2255343
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.14:93207589 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AGTCTGCCGCTTCCTCACCCCAAGC[C/T]CACCCTTCCCTTTTAACCCCAACCT
Phenotype
MIM: 613816
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GON7 PubMed Links

Gene Details

Gene
GON7
Gene Name
GON7, KEOPS complex subunit homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032490.4 Intron NP_115879.2
Gene
UBR7
Gene Name
ubiquitin protein ligase E3 component n-recognin 7 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175748.3 Intron NP_786924.2

View Full Product Details