Product Details

SNP ID
rs8135177
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:42561282 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCGTCGGATGGAAAATGTGCTAAG[C/T]TGGGCAACACAACTTTGCTTCCTGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SERHL2 PubMed Links

Gene Details

Gene
SERHL2
Gene Name
serine hydrolase-like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284334.1 Intron NP_001271263.1
NM_014509.4 Intron NP_055324.2
XM_017028738.1 Intron XP_016884227.1
XM_017028739.1 Intron XP_016884228.1

View Full Product Details